Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bet bromodomain inhibitor 4 | 2407658-41-5 | ≥98.0% | 518.59 g/mol | C27H31FN8O2 | 10 MG
SDP

Catalog No. 5000287924
Encompass_Preferred

BET bromodomain inhibitor 4 is a small-molecule inhibitor of BET family bromodomain epigenetic reader domains supplied for research use as a 10 mg powder. It is used in preclinical studies to probe BET-mediated epigenetic regulation and assay bromodomain interactions.

  • Inhibits BET bromodomain activity.
  • Intended for research use only.
  • Supplied as a 10 mg powder for laboratory applications.
  • Target: epigenetic reader domain (BET bromodomain).
  • CAS number: 2407658-41-5.
  • Molecular formula: C27H31FN8O2; molecular weight: 518.59 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-87924 Supplier Medchemexpress LLC Supplier No. HY15739310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BET bromodomain inhibitor 4 is a small-molecule inhibitor of BET family bromodomain epigenetic reader domains supplied for research use as a 10 mg powder. It is used in preclinical studies to probe BET-mediated epigenetic regulation and assay bromodomain interactions.

  • Inhibits BET bromodomain activity.
  • Intended for research use only.
  • Supplied as a 10 mg powder for laboratory applications.
  • Target: epigenetic reader domain (BET bromodomain).
  • CAS number: 2407658-41-5.
  • Molecular formula: C27H31FN8O2; molecular weight: 518.59 g/mol.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.