Learn More
Medchemexpress LLC BET-IN-14 | 2243669-93-2 | C30H37N7O2 | 1 MG
SDP
Catalog No. 5000373510
Encompass_Preferred
BET-IN-14 is an orally active pan BET inhibitor (IC50: 5.35 nM) exhibiting anti-cancer activity. It shows anti-tumor effects against various cancer cell lines, including MM.1S, Ty-82, MV-4-11, and KG-1 cells, with an average IC50 of 174.7 nM after 72 hours. In vivo, BET-IN-14 inhibits tumor growth in MV-4-11 xenograft models.
  • Orally active pan BET inhibitor
  • Exhibits anti-cancer activity
  • Effective against various cancer cell lines (MM.1S, Ty-82, MV-4-11, KG-1)
  • Inhibits tumor growth in xenograft models
Catalog No. 50-003-73510 Supplier Medchemexpress LLC Supplier No. HY1532261MG
BET-IN-14 is an orally active pan BET inhibitor (IC50: 5.35 nM) exhibiting anti-cancer activity. It shows anti-tumor effects against various cancer cell lines, including MM.1S, Ty-82, MV-4-11, and KG-1 cells, with an average IC50 of 174.7 nM after 72 hours. In vivo, BET-IN-14 inhibits tumor growth in MV-4-11 xenograft models.
  • Orally active pan BET inhibitor
  • Exhibits anti-cancer activity
  • Effective against various cancer cell lines (MM.1S, Ty-82, MV-4-11, KG-1)
  • Inhibits tumor growth in xenograft models