Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BET-IN-14 | 2243669-93-2 | C30H37N7O2 | 25 MG
SDP

Catalog No. 5000375622
Encompass_Preferred

BET-IN-14 is an orally active pan BET inhibitor with an IC50 of 5.35 nM. It exhibits anti-cancer activity. In vitro studies show that BET-IN-14 (Compound 171) has anti-tumor activity against various cancer cell lines (average IC50: 174.7 nM), particularly in MM.1S, Ty-82, MV-4-11, and KG-1 cells. In vivo, BET-IN-14 (50 and 100 mg/kg, p.o., qd) inhibits tumor growth in MV-4-11 xenograft models by 60.36%.

  • Orally active pan BET inhibitor with high potency (IC50 of 5.35 nM)
  • Displays anti-tumor activity against various cancer cell lines (average IC50: 174.7 nM)
  • Effective in MM.1S, Ty-82, MV-4-11, and KG-1 cells
  • Inhibits tumor growth in MV-4-11 xenograft models by over 60% at tested dosages

Catalog No. 50-003-75622 Supplier Medchemexpress LLC Supplier No. 25MGHY153226
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BET-IN-14 is an orally active pan BET inhibitor with an IC50 of 5.35 nM. It exhibits anti-cancer activity. In vitro studies show that BET-IN-14 (Compound 171) has anti-tumor activity against various cancer cell lines (average IC50: 174.7 nM), particularly in MM.1S, Ty-82, MV-4-11, and KG-1 cells. In vivo, BET-IN-14 (50 and 100 mg/kg, p.o., qd) inhibits tumor growth in MV-4-11 xenograft models by 60.36%.

  • Orally active pan BET inhibitor with high potency (IC50 of 5.35 nM)
  • Displays anti-tumor activity against various cancer cell lines (average IC50: 174.7 nM)
  • Effective in MM.1S, Ty-82, MV-4-11, and KG-1 cells
  • Inhibits tumor growth in MV-4-11 xenograft models by over 60% at tested dosages

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.