Learn More
Medchemexpress LLC Bet-in-19 | 1643947-30-1 | 99.7% | 333.39 | C19H19N5O | 10MG
SDP
Catalog No. 5000238326
Encompass_Preferred
BET-IN-19 is a small-molecule BET bromodomain inhibitor provided as a high-purity research reagent for epigenetic and oncology studies. It is supplied as a powder in multiple pack sizes and displays cellular activity against AML MV4-11 cells, supporting biochemical and cell-based assays. Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • High purity (99.74%).
  • Molecular formula C19H19N5O; molecular weight 333.39.
  • Demonstrated inhibition of hIL-6 transcription and c-myc activity (IC50 ≤ 0.3 μM).
  • Available in multiple pack sizes for flexible experimental design.
  • Supplied as a powder with defined cold-storage stability.
Catalog No. 50-002-38326 Supplier Medchemexpress LLC Supplier No. HY12523610MG
BET-IN-19 is a small-molecule BET bromodomain inhibitor provided as a high-purity research reagent for epigenetic and oncology studies. It is supplied as a powder in multiple pack sizes and displays cellular activity against AML MV4-11 cells, supporting biochemical and cell-based assays. Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • High purity (99.74%).
  • Molecular formula C19H19N5O; molecular weight 333.39.
  • Demonstrated inhibition of hIL-6 transcription and c-myc activity (IC50 ≤ 0.3 μM).
  • Available in multiple pack sizes for flexible experimental design.
  • Supplied as a powder with defined cold-storage stability.