Learn More
Medchemexpress LLC Beta-1 | 2924598-24-1 | C37H28IN3S2 | 25 MG
SDP
Catalog No. 5000374376
Encompass_Preferred
BETA-1 is the first twisted intramolecular charge transfer (TICT)-aggregation-induced emission (AIE) integration molecule. It emits cyan fluorescence in lipid droplets (LDs) and red fluorescence in mitochondria. This molecule can be used for the simultaneous and dual-color imaging of LDs and mitochondria in vivo and in vitro.
  • Simultaneous and dual-color imaging of lipid droplets and mitochondria in vivo and in vitro
  • Suitable for imaging of lipid droplets and mitochondria in live cells
  • Emits cyan fluorescence with excitation at 405 nm and emission at 430-500 nm
  • Emits red fluorescence with excitation at 561 nm and emission at 600-700 nm
Catalog No. 50-003-74376 Supplier Medchemexpress LLC Supplier No. 25MGHY152073
BETA-1 is the first twisted intramolecular charge transfer (TICT)-aggregation-induced emission (AIE) integration molecule. It emits cyan fluorescence in lipid droplets (LDs) and red fluorescence in mitochondria. This molecule can be used for the simultaneous and dual-color imaging of LDs and mitochondria in vivo and in vitro.
  • Simultaneous and dual-color imaging of lipid droplets and mitochondria in vivo and in vitro
  • Suitable for imaging of lipid droplets and mitochondria in live cells
  • Emits cyan fluorescence with excitation at 405 nm and emission at 430-500 nm
  • Emits red fluorescence with excitation at 561 nm and emission at 600-700 nm