Learn More
Medchemexpress LLC BETA-1 | 2924598-24-1 | C37H28IN3S2 | 5 MG
SDP
Catalog No. 5000374132
Encompass_Preferred
BETA-1 is the first twisted intramolecular charge transfer (TICT)-aggregation-induced emission (AIE) integration molecule. It emits cyan fluorescence in lipid droplets (LDs) and red fluorescence in mitochondria. BETA-1 can be utilized for simultaneous and dual-color imaging of LDs and mitochondria both in vivo and in vitro.
  • Emits cyan fluorescence in lipid droplets (LDs).
  • Emits red fluorescence in mitochondria.
  • Can be used for simultaneous and dual-color imaging of LDs and mitochondria in vivo and in vitro.
Catalog No. 50-003-74132 Supplier Medchemexpress LLC Supplier No. HY1520735MG
BETA-1 is the first twisted intramolecular charge transfer (TICT)-aggregation-induced emission (AIE) integration molecule. It emits cyan fluorescence in lipid droplets (LDs) and red fluorescence in mitochondria. BETA-1 can be utilized for simultaneous and dual-color imaging of LDs and mitochondria both in vivo and in vitro.
  • Emits cyan fluorescence in lipid droplets (LDs).
  • Emits red fluorescence in mitochondria.
  • Can be used for simultaneous and dual-color imaging of LDs and mitochondria in vivo and in vitro.