Learn More
New England Biolabs, Inc. β1-3,4 Galactosidase - 400 units
SDP
Catalog No. NC1207877
Encompass
b1-3,4 Galactosidase is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b1-3 and b1-4-linked galactose residues from oligosaccharides.
Catalog No. NC1207877 Supplier New England Biolabs, Inc. Supplier No. P0746S
b1-3,4 Galactosidase is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b1-3 and b1-4-linked galactose residues from oligosaccharides.