Learn More
New England Biolabs, Inc. β1-3 Galactosidase - 500 units
SDP
Catalog No. 50811840
Encompass
b1-3 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-3 and, at a much lower rate, 1-6 linked galactose residues from oligosaccharides.
Catalog No. 50-811-840 Supplier New England Biolabs, Inc. Supplier No. P0726S
b1-3 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-3 and, at a much lower rate, 1-6 linked galactose residues from oligosaccharides.