Learn More
New England Biolabs, Inc. β1-4 Galactosidase S - 2000 units
SDP
Catalog No. 501047515
Encompass
b1-4 Galactosidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b1-4 linked galactose residues from oligosaccharides.
Catalog No. 50-104-7515 Supplier New England Biolabs, Inc. Supplier No. P0745L
b1-4 Galactosidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing b1-4 linked galactose residues from oligosaccharides.