Learn More
Sigma Aldrich Fine Chemicals Biosciences Beta 14 galactosyltransfer
Catalog No. 502905036
Encompass
Recombinant human Beta-1 4-galactosyltransferase 1 (B4GALT1) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 40 kDa. The DTT-reduced protein migrates as a ~55 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein.
Catalog No. 50-290-5036 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE009350UG
Recombinant human Beta-1 4-galactosyltransferase 1 (B4GALT1) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 40 kDa. The DTT-reduced protein migrates as a ~55 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein.