Learn More
ABclonal Technology beta 2 Microglobulin
SDP
Supplier:  ABclonal Technology A23430100UL
Encompass_Preferred
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions The encoded antimicrobial protein displays antibacterial activity in amniotic fluid A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia
Catalog No. 50-000-05242