Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ beta 2-Microglobulin Antibody Pair [HRP]
SDP

Catalog No. NB009797
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009797 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009797 Supplier Novus Biologicals™ Supplier No. NBP27937915PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 3.13 pg/mL
Assay Sensitivity 3.13-200 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human beta 2-Microglobulin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Gene ID (Entrez) 567
Gene Symbol B2M
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human beta 2-Microglobulin monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of mouse anti-Human beta 2-Microglobulin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 24 ng of recombinant Human beta 2-Microglobulin.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
    Storage Requirements Storage is content dependent.
    Target beta 2-Microglobulin
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 200 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.