Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beta-2-microglobulin (B2M) Human ProcartaPlex™ Simplex Kit

Catalog No. 502365747
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-236-5747 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-236-5747 Supplier Invitrogen™ Supplier No. EPX01012414901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Human Beta-2-microglobulin (B2M) Simplex ProcartaPlex Kit measures beta-2-microglobulin (B2M) protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

  • The Human Beta-2-microglobulin (B2M) Simplex ProcartaPlex Kit measures beta-2-microglobulin (B2M) protein
  • Designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation
  • When combining multiple Simplex kits (e.g., when not using a pre-configured multiplex panel), only one buffer kit (EPX010-10420-901) is needed for each assay plate regardless of plex size
  • Recommended sample pre-dilution for serum and plasma samples is 1:100

About ProcartaPlex assays for the Luminex platform

  • ProcartaPlex immunoassays are based on the principles of a sandwich ELISA
  • They use two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument
  • ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant
  • Analyzed results can be obtained in just four hours
  • Validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • Panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine)
  • Luminex technology is the most referenced multiplexing platform for protein detection and quantitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Region 21
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Simplex Kit
For Use With (Equipment) Luminex Instruments
Gene Beta-2-microglobulin
Gene Alias IMD43
Gene ID (Entrez) 567
Gene Symbol B2M
Interassay CV Less than 15%
Intraassay CV Less than 15%
Product Line ProcartaPlex
Protein Beta-2-microglobulin
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
UniProt ID P61769
Content And Storage

• 1 vial Capture Beads (50X)
• 1 vial Biotinylated Detection Antibody (50X)
• 2 vials Human Standard Mix 8 (lyophilized)
• 1 Bottle Universal Assay Buffer (10X)

Store at 2°C to 8°C.

Research Area Oncology, Toxicology
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.