Learn More
Drg International Inc Beta-2 Microglobulin Elisa
Supplier:  Drg International Inc EIA1789
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of beta-2 microglobulin (B2MG) concentration in serum. Beta-2-microglobulin (Beta2-MG) is expressed by the nucleated cells of the body and on many tumor lines. The Beta2-MG ELISA provides a sensitive and reliable assay for the measurement of Beta2-microglobulin in human serum. The Beta2-MG ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay (ELISA). The assay system utilizes a monoclonal anti-Beta2-MG antibody for solid phase immobilization (on the microtiter wells). A sheep anti-Beta2-MG antibody is in the antibody-enzyme (horseradish peroxidase) conjugate solution.
Catalog No. 50-194-7542