Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-(4-Pyridyl)acrolein Oxalate 98.0+%, TCI America™
SDP

Catalog No. P11481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11481G Supplier TCI America Supplier No. P11481G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 383177-50-2
Molecular Formula C10H9NO5
Molecular Weight (g/mol) 223.18
MDL Number MFCD00191489
InChI Key NSTHNVMITOXEJZ-UHFFFAOYSA-N
PubChem CID 21974133
IUPAC Name 3-(pyridin-4-yl)prop-2-enal; oxalic acid
SMILES OC(=O)C(O)=O.O=CC=CC1=CC=NC=C1

Specifications

Melting Point 180°C
Color Yellow
Quantity 1 g
Formula Weight 223.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-(4-Pyridyl)acrolein Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.