Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-Acetoxyisovalerylshikonin | 69091-17-4 | 25 MG
SDP

Catalog No. 5000421914
Encompass_Preferred

Beta-Acetoxyisovalerylshikonin is a naphthoquinone derivative isolated from Arnebia euchroma.

  • High purity: 99.57%
  • Molecular weight: 430.45
  • Molecular formula: C23H26O8
  • Appearance: Brown to black solid
  • Used in simultaneous densitometric determination of related compounds
  • Applied in isolation and purification processes using preparative HPLC

Catalog No. 50-004-21914 Supplier Medchemexpress LLC Supplier No. 25MGHYN2188
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta-Acetoxyisovalerylshikonin is a naphthoquinone derivative isolated from Arnebia euchroma.

  • High purity: 99.57%
  • Molecular weight: 430.45
  • Molecular formula: C23H26O8
  • Appearance: Brown to black solid
  • Used in simultaneous densitometric determination of related compounds
  • Applied in isolation and purification processes using preparative HPLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.