Learn More
enQuire Bioreagents Anti-beta Actin antibody | 50ug
Supplier:  enQuire Bioreagents AB822750UG
Encompass_Preferred
Rabbit polyclonal to beta Actin. The immunogen used for this product shares 77% homology with Gamma actin/actin cytoplasmic 2. Cross-reactivity with this protein has not been confirmed experimentally. Reacts with: Mouse, Rat, Sheep, Rabbit, Chicken, Cow, Dog, Human, Pig, Xenopus laevis, Fish, Monkey, Rhesus monkey, Chinese hamster.
Catalog No. NC1666557