Learn More
Santa Cruz Biotechnology Anti-beta-Actin Antibody (C4) HRP, 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC47778 HRP
Encompass
Anti-beta-Actin Antibody (C4) is recommended for detection of beta-Actin of mouse, rat, human, avian, bovine, canine, porcine, rabbit, Dictyostelium discoideum and Physarum polycephalum origin by WB, IP, IF, IHC(P) and ELISA; may cross-react with all six known isoforms of Actin in higher vertebrates (including cytoplasmic beta- and gamma- Actin isoforms, skeletal, cardiac, and vascular alpha-Actin isoforms, and enteric gamma-Actin isoform).
Catalog No. NC0280063