Learn More
Bioss Beta Actin CAEEL/Loading Contr
SDP
Catalog No. 5000064878
Encompass_Preferred
Beta Actin (CAEEL Loading Control) Primary Antibody Unconjugated Host Species Mouse Target Species Caenorhabditis Elegans Clonality Monoclonal Applications WB Concentration 1ug/ul Size 100 ul Storage Buffer 0 01M TBS(pH7 4) with 1% BSA 0 02% Proclin300 and 50% Glycerol
Catalog No. 50-000-64878 Supplier Bioss Supplier No. BSM8770M
Beta Actin (CAEEL Loading Control) Primary Antibody Unconjugated Host Species Mouse Target Species Caenorhabditis Elegans Clonality Monoclonal Applications WB Concentration 1ug/ul Size 100 ul Storage Buffer 0 01M TBS(pH7 4) with 1% BSA 0 02% Proclin300 and 50% Glycerol