Learn More
Medchemexpress LLC Beta Actin (Fruit Fly) Antibody | 10 UL
SDP
Catalog No. 5000440782
Encompass_Preferred
Beta Actin (Fruit Fly) Antibody is a non-conjugated and Rabbit origined IgG polyclonal antibody, targeting to Beta Actin (Fruit Fly). It can be used as a loading control antibody.
  • Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
  • Multiple isoforms are involved in various cellular functions such as cytoskeleton structure, cell mobility, chromosome movement and muscle contraction.
  • Subcellular Localization: Cytoplasm, cytoskeleton.
  • Subunit: Interacts with Rab6.
Catalog No. 50-004-40782 Supplier Medchemexpress LLC Supplier No. HYP8125410UL
Beta Actin (Fruit Fly) Antibody is a non-conjugated and Rabbit origined IgG polyclonal antibody, targeting to Beta Actin (Fruit Fly). It can be used as a loading control antibody.
  • Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
  • Multiple isoforms are involved in various cellular functions such as cytoskeleton structure, cell mobility, chromosome movement and muscle contraction.
  • Subcellular Localization: Cytoplasm, cytoskeleton.
  • Subunit: Interacts with Rab6.