Learn More
Enzo Life Sciences beta-Actin polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905733100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human β-Actin. UniProt ID: P60709. Species reactivity: Human, Mouse, Rat. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (10µg/ml) Western Blot (1-2µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7023