Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Alaninamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A13915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13915G 5 g
A13911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13915G Supplier TCI America Supplier No. A13915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64017-81-8 | C3H9ClN2O | 124.568 g/mol

Chemical Identifiers

CAS 64017-81-8
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.568
MDL Number MFCD00060747
InChI Key GAGJMOQGABUOBK-UHFFFAOYSA-N
Synonym 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl
PubChem CID 22222288
IUPAC Name 3-aminopropanamide;hydrochloride
SMILES C(CN)C(=O)N.Cl

Specifications

Melting Point 153°C
Color White
Quantity 5 g
Formula Weight 124.57
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material beta-Alaninamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.