Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Alanine, Spectrum™ Chemical
SDP

Catalog No. 18620174 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-174 100 g
18-620-175 500 g
18-620-176 2.5 kg
18-620-177 12 kg
18-620-178 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-174 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL120100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-95-9

Specifications

Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 101%
Grade Reagent
Chemical Name or Material Beta-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.