Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Alanyl-L-histidine, 95%

Catalog No. AAH6566814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6566814 25 g
AAH6566806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6566814 Supplier Thermo Scientific Chemicals Supplier No. H6566814

CAS: 305-84-0 | C9H14N4O3 | 226.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-84-0
Molecular Formula C9H14N4O3
Molecular Weight (g/mol) 226.236
MDL Number MFCD00005207
InChI Key CQOVPNPJLQNMDC-ZETCQYMHSA-N
Synonym l-carnosine, carnosine, beta-alanyl-l-histidine, karnozin, karnozzn, ignotine, l-ignotine, n-2-m, unii-8ho6pvn24w, nalpha-beta-alanyl-l-histidine
PubChem CID 439224
ChEBI CHEBI:15727
IUPAC Name (2S)-2-(3-aminopropanoylamino)-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)NC(=O)CCN

Specifications

Melting Point 253°C
Color White
Beilstein 87671
Quantity 25 g
Formula Weight 226.24
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material beta-Alanyl-L-histidine
EINECSNumber 206-169-9
RTECSNumber MS3080000
TSCA Yes
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.