Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Amylase from sweet po
Catalog No. 502907009
Encompass
-Amylase from sweet potato is a tetrameric enzyme with each monomer comprising (Î/) core domain and a C-terminal loop region. It belongs to the glycoside hydrolase family GH14.
Catalog No. 50-290-7009 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A87815X1VL
-Amylase from sweet potato is a tetrameric enzyme with each monomer comprising (Î/) core domain and a C-terminal loop region. It belongs to the glycoside hydrolase family GH14.