Learn More
CPC Scientific H-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD019A
Encompass
Amyloid fragment shown to produce both the neurotoxic and the neurotrophic effects of beta-amyloid protein on cultured neurons. Furthermore, the peptide forms aggregates and typical amyloid fibrils resembling those of the beta-amyloid protein in senile plaque cores.ONE-LETTER SEQUENCE: EDVGSNKGAIIGLMMOLECULAR FORMULA: C59H102N16O21S1MOLECULAR WEIGHT:1403.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: 144189-71-9, SYNONYMS: Amyloid β-Protein (22-35)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: T. Takadera et al., Neurosci. Letters, 161, 41 (1993)
Catalog No. 50-210-8672