Learn More
Apexbio Technology LLC Beta-Amyloid (1-11) 190436-05-6 25mg
SDP
Catalog No. 5000092826
Encompass_Preferred
Beta-Amyloid (1-11) (CAS 190436-05-6) is a synthetic peptide fragment derived from the N-terminus of the amyloid precursor protein comprising the amino acid sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-OH This peptide represents a portion of the beta-amyloid (A ) sequence implicated in amyloidogenic pathways associated with neurodegenerative processes particularly Alzheimer s disease Beta-Amyloid (1-11) is utilized in research to investigate amyloid aggregation dynamics immunogenic epitope mapping within the A region and the biochemical properties of amyloidogenic peptides It serves as a molecular tool in studies examining the early stages of A processing clearance and interactions with cellular components
Catalog No. 50-000-92826 Supplier Apexbio Technology LLC Supplier No. A100225
Beta-Amyloid (1-11) (CAS 190436-05-6) is a synthetic peptide fragment derived from the N-terminus of the amyloid precursor protein comprising the amino acid sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-OH This peptide represents a portion of the beta-amyloid (A ) sequence implicated in amyloidogenic pathways associated with neurodegenerative processes particularly Alzheimer s disease Beta-Amyloid (1-11) is utilized in research to investigate amyloid aggregation dynamics immunogenic epitope mapping within the A region and the biochemical properties of amyloidogenic peptides It serves as a molecular tool in studies examining the early stages of A processing clearance and interactions with cellular components