Learn More
Echelon Biosciences Research Labs Beta Amyloid (1-16), rat - 5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 642365MG
Encompass_Preferred
Beta Amyloid (1-16), rat - 5mg , Asp-Ala-Glu-Phe-Gly-His-Asp-Ser-Gly-Phe-Glu-Val-Arg-His-Gln-Lys-OH Category/Application: Alzheimer's This item is non returnable
Catalog No. 50-190-8769