Learn More
Enzo Life Sciences beta-Amyloid (1-40) (1mg). CAS: 131438-79-4
SDP
Supplier:  Enzo Life Sciences ALX151003P001
Encompass_Preferred
Formula: C194H295N53O58S. MW: 4329.9. Purity: ≥95%. Solubility: Soluble in 5% acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-7752