Learn More
Medchemexpress LLC beta Amyloid 1-40 Antibody (YA3554) | 10 UL
SDP
Catalog No. 5000443433
Encompass_Preferred
beta Amyloid 1-40 Antibody (YA3554) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting beta Amyloid 1-40. It shows reactivity with Human and Mouse samples. The antibody is supplied as a liquid in PBS (pH7.4) with 0.1% BSA, 40% Glycerol, and 0.05% Sodium Azide as a preservative. It is affinity purified.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets beta Amyloid 1-40
  • Reacts with human and mouse samples
  • Supplied in PBS (pH7.4), 0.1% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Affinity purified
  • Applications include WB (1:1000), IHC-P (1:1000), and IF-Tissue (1:200)
Catalog No. 50-004-43433 Supplier Medchemexpress LLC Supplier No. HYP8375310UL
beta Amyloid 1-40 Antibody (YA3554) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting beta Amyloid 1-40. It shows reactivity with Human and Mouse samples. The antibody is supplied as a liquid in PBS (pH7.4) with 0.1% BSA, 40% Glycerol, and 0.05% Sodium Azide as a preservative. It is affinity purified.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets beta Amyloid 1-40
  • Reacts with human and mouse samples
  • Supplied in PBS (pH7.4), 0.1% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Affinity purified
  • Applications include WB (1:1000), IHC-P (1:1000), and IF-Tissue (1:200)