Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta Amyloid 1-40 Antibody (YA3554) | 50 UL
SDP

Catalog No. 5000444165
Encompass_Preferred

The beta Amyloid 1-40 Antibody (YA3554) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target beta Amyloid 1-40. This affinity-purified antibody is supplied as a liquid in PBS (pH 7.4) with 0.1% BSA, 40% Glycerol, and 0.05% Sodium Azide. It is suitable for a range of research applications, including Western Blotting (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunofluorescence-Tissue (IF-Tissue).

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Conjugation: Non-conjugated
  • Molecular weight: 4 kDa
  • Formulation: Supplied in PBS (pH7.4), 0.1% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage and stability: Store at -20°C for 1 year; avoid repeated freeze / thaw cycles
  • Appearance: Liquid
  • Shipping: With blue ice
  • Application dilution ratios: WB: 1:1000, IHC-P: 1:1000, IF-Tissue: 1:200

Catalog No. 50-004-44165 Supplier Medchemexpress LLC Supplier No. HYP8375350UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The beta Amyloid 1-40 Antibody (YA3554) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target beta Amyloid 1-40. This affinity-purified antibody is supplied as a liquid in PBS (pH 7.4) with 0.1% BSA, 40% Glycerol, and 0.05% Sodium Azide. It is suitable for a range of research applications, including Western Blotting (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunofluorescence-Tissue (IF-Tissue).

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Conjugation: Non-conjugated
  • Molecular weight: 4 kDa
  • Formulation: Supplied in PBS (pH7.4), 0.1% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage and stability: Store at -20°C for 1 year; avoid repeated freeze / thaw cycles
  • Appearance: Liquid
  • Shipping: With blue ice
  • Application dilution ratios: WB: 1:1000, IHC-P: 1:1000, IF-Tissue: 1:200

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.