Learn More
Enzo Life Sciences beta-Amyloid (1-42) (1mg). CAS: 107761-42-2
SDP
Supplier:  Enzo Life Sciences ALX151002P001
Encompass_Preferred
Formula: C203H311N55O60S. MW: 4514.1. Purity: ≥95% (HPLC). Appearance: White to off-white lyophilized powder. Solubility: Soluble in 5% acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-7750