Learn More
CPC Scientific H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD023B
Encompass
Similar to the beta-amyloid protein fragment (25-35), the fragment (1-38) destabilizes calcium homeostasis and makes neurons vulnerable to environmental insults.ONE-LETTER SEQUENCE: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGMOLECULAR FORMULA: C184H277N51O56S1MOLECULAR WEIGHT:4131.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [131438-74-9], SYNONYMS: Amyloid β-Protein (1-38)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: M.P. Mattson, J. Neurosci., 12, 376 (1992)
Catalog No. 50-210-8675