Learn More
Enzo Life Sciences beta-Amyloid (25-35) (1mg). CAS: 131602-53-4
SDP
Supplier:  Enzo Life Sciences ALX151008P001
Encompass_Preferred
Formula: C45H81N13O14S. MW: 1060.3. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7755