Learn More
Enzo Life Sciences beta-Amyloid (25-35) (5mg). CAS: 131602-53-4
SDP
Supplier:  Enzo Life Sciences ALX151008P005
Encompass_Preferred
Formula: C45H81N13O14S. MW: 1060.3. Purity: ≥99% (HPLC). Appearance: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7756