Learn More
TARGETMOL CHEMICALS INC beta-Amyloid 31-35 10MG
SDP
Catalog No. 502878014
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. beta-Amyloid (31-35) (beta-Amyloid 31-35) is the shortest sequence of native Amyloid-beta peptide. Purity 100%
Catalog No. 50-287-8014 Supplier TARGETMOL CHEMICALS INC Supplier No. T756210MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. beta-Amyloid (31-35) (beta-Amyloid 31-35) is the shortest sequence of native Amyloid-beta peptide. Purity 100%