Learn More
CPC Scientific Ala-Lys-Glu-Arg-Leu-Glu-Ala-Lys-His-Arg-Glu-Arg-Met-Ser-Gln-Val-Met-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD029B
Encompass
17-mer peptide that reduces the neurologic damage in vivo in a model of central nervous system ischemia, possibly through its effect on synaptic plasticity.ONE-LETTER SEQUENCE: AKERLEAKHRERMSQVMMOLECULAR FORMULA: C86H151N31O26S2MOLECULAR WEIGHT:2099.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: H. Ninomiya et al., J. Cell Biol., 121, 879 (1993); M.P. Bowes et al., Exp. Neurol., 129, 112 (1994)
Catalog No. 50-210-8687