Learn More
R Peptide LLC Beta Amyloid (1-42) Aggregation Kit, 0.25 mg, White lyophilized powder, -20°C storage

Supplier: R Peptide LLC A1170025
The R PEPTIDE LLC Beta Amyloid (1-42) Aggregation Kit is designed for researchers studying the aggregation kinetics of Beta-Amyloid (1-42) peptide, which is linked to Alzheimer's Disease. This comprehensive kit ensures high purity and batch consistency, featuring a thioflavin fluorescence assay for effective monitoring of aggregation dynamics. Ideal for use in neurodegenerative disease research, the kit is produced using recombinant technology in *E. coli* and includes the necessary components for detailed studies on aggregate stability and potential therapeutic interventions.
- Includes Beta-Amyloid (1-42) peptide
- Produced using recombinant technology
- High purity over 97% as per mass spec
- Thioflavin fluorescence assay for detection
- Suitable for aggregation studies and Alzheimer's research
- White lyophilized powder
- Store at -20°C
- Ambient shipping conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.