Learn More
CPC Scientific H-Asp-Ala-Glu-Phe-Gly-His-Asp-Ser-Gly-Phe-Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD017B
Encompass
ONE-LETTER SEQUENCE: DAEFGHDSGFEVRHQKLVFFAEDVGSNKGAIIGLMVGGVVIAMOLECULAR FORMULA: C199H307N53O59S1MOLECULAR WEIGHT:4418STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [107761-42-2], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: E. Paradis et al., J. Neurosci., 16, 7533 (1996); J. Murrell et al., Science, 254, 97 (1991); D. Goldgabber et al., Science, 235, 877 (1987)
Catalog No. 50-210-8669