Learn More
CPC Scientific H-Asp-Ala-Glu-Phe-Gly-His-Asp-Ser-Gly-Phe-Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AMYD018B
Encompass
ONE-LETTER SEQUENCE: DAEFGHDSGFEVRHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C190H291N51O57S1MOLECULAR WEIGHT:4233.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [131438-79-4], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: A. Klegeris et al., Biochem. Biophys. Res. Commun., 199, 984 (1994); M.A.Kowalska et al., Biochem. Biophys. Res. Commun., 205, 1829 (1994); B.A. Yankner et al., Science, 250, 279 (1990)
Catalog No. 50-210-8671