Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals beta-Apo-oxytetracycline, 'can be used as secondary standard'

Catalog No. AC257731000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257731000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257731000 Supplier Thermo Scientific Chemicals Supplier No. 257731000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Beta-apo-oxytetracycline, CAS # 18751-99-0, is a member of the tetracycline family and is a degradation production formed from oxytetracycline. This compound displays antibiotic activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Beta-apo-oxytetracycline is a degradation product derived from oxytetracycline and displays antibiotic activity
• Similar to its parent compound, oxytetracycline, beta-apo-oxytetracycline can also inhibit protein synthesis by binding to the 30S ribosomal subunit

Applications

• In laboratory assays, it promotes the inhibition of the growth of aerobic sludge bacteria, Pseudomonas, Agrobacterium, Moraxella, Bacillus, as well as tetracycline-resistant strains of E. coli

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18751-99-0
Molecular Formula C22H22N2O8
Molecular Weight (g/mol) 442.39
MDL Number MFCD00062827
InChI Key DRKMHDAKULCOKQ-UHFFFAOYSA-N
Synonym beta-apo-oxytetracycline, alpha-apo-oxytetracycline, alpha-apooxytetracycline, alpha-apoterramycin, beta-apo oxytetracycline, 2z-2-amino hydroxy methylidene-5-4,5-dihydroxy-9-methyl-3-oxo-1h-benzo f 2 benzofuran-1-yl-4-dimethylamino-6-hydroxycyclohexane-1,3-dione, 4-1,3-dihydro-4,5-dihydroxy-9-methyl-3-oxonaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxo-1-cyclohexene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1,3-dihydronaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxohydrobenzo f isobenzofuranyl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-enecarboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1h-naphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
PubChem CID 54710410
IUPAC Name 4-(4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl)-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
SMILES CC1=C2C(OC(=O)C2=C(C3=C1C=CC=C3O)O)C4C(C(=C(C(=O)C4O)C(=O)N)O)N(C)C

Specifications

Melting Point 200.0°C to 204.0°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 100 mg
Merck Index 10, 6846
Specific Rotation − 207.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution and ammonia
Formula Weight 442.39
Percent Purity ≥97.0% (anhydrous and solvent free), ≥80 wt% (as is, calculated)(HPLC)
Physical Form Powder
Chemical Name or Material β-Apo-oxytetracycline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.