Learn More
TARGETMOL CHEMICALS INC beta-Asarone 10MG
SDP
Catalog No. 502876674
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. beta-Asarone (Cis-Isoelemicin) may be a potential preventive drug for Alzheimer's disease. 2. Beta-Asarone prevents autophagy and synaptic loss by reducing ROCK expression in SAMP8 mice. 3. The co-administration of beta-Asarone(Cis-Isoasarone) and l-dopa may contribute to the treatment of 6-OHDA-induced damage in rats by inhibiting autophagy activity. 4. Beta-Asarone exhibits anti-inflammatory effects by suppressing the production of pro-inflammatory mediators through NF-kB signaling and the JNK pathways in activated microglial cells and might be developed as a promising candidate to treat various neuroinflammatory diseases. Purity 99.32%
Catalog No. 50-287-6674 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S196210MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. beta-Asarone (Cis-Isoelemicin) may be a potential preventive drug for Alzheimer's disease. 2. Beta-Asarone prevents autophagy and synaptic loss by reducing ROCK expression in SAMP8 mice. 3. The co-administration of beta-Asarone(Cis-Isoasarone) and l-dopa may contribute to the treatment of 6-OHDA-induced damage in rats by inhibiting autophagy activity. 4. Beta-Asarone exhibits anti-inflammatory effects by suppressing the production of pro-inflammatory mediators through NF-kB signaling and the JNK pathways in activated microglial cells and might be developed as a promising candidate to treat various neuroinflammatory diseases. Purity 99.32%