Learn More
Medchemexpress LLC Beta-asarone | 5273-86-9 | 208.25 | 25 MG
SDP
Catalog No. 5000390265
Encompass_Preferred
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent. It is the major ingredient of Acorus tatarinowii Schott. It can protect nerve cells from apoptosis and autophagy, inhibit the expression of α-synuclein, and provide myocardial protection, making it suitable for the study of neurological and cardiovascular diseases.
  • Orally active and BBB-penetrable
  • Anti-inflammatory agent
  • Neuroprotective agent
  • Protects nerve cells from apoptosis and autophagy
  • Inhibits expression of α-synuclein
  • Provides myocardial protection
  • Used in the study of neurological and cardiovascular diseases
Catalog No. 50-003-90265 Supplier Medchemexpress LLC Supplier No. 25MGHYN1501
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent. It is the major ingredient of Acorus tatarinowii Schott. It can protect nerve cells from apoptosis and autophagy, inhibit the expression of α-synuclein, and provide myocardial protection, making it suitable for the study of neurological and cardiovascular diseases.
  • Orally active and BBB-penetrable
  • Anti-inflammatory agent
  • Neuroprotective agent
  • Protects nerve cells from apoptosis and autophagy
  • Inhibits expression of α-synuclein
  • Provides myocardial protection
  • Used in the study of neurological and cardiovascular diseases