Learn More
Medchemexpress LLC Beta-asarone | 5273-86-9 | MFCD00009281 | 208.25 | 1 ML
SDP
Catalog No. 5000415648
Encompass_Preferred
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent, found as a major ingredient in *Acorus tatarinowii Schott*. It can protect nerve cells from apoptosis and autophagy, inhibit the expression of α-synuclein, and offers myocardial protection. This product can be used in the study of neurological and cardiovascular diseases.
  • Orally active and blood-brain barrier (BBB)-penetrable.
  • Anti-inflammatory properties.
  • Neuroprotective agent.
  • Protects nerve cells from apoptosis and autophagy.
  • Inhibits expression of α-synuclein.
  • Provides myocardial protection.
  • Useful for research in neurological and cardiovascular diseases.
Catalog No. 50-004-15648 Supplier Medchemexpress LLC Supplier No. HYN150110MM/1ML
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent, found as a major ingredient in *Acorus tatarinowii Schott*. It can protect nerve cells from apoptosis and autophagy, inhibit the expression of α-synuclein, and offers myocardial protection. This product can be used in the study of neurological and cardiovascular diseases.
  • Orally active and blood-brain barrier (BBB)-penetrable.
  • Anti-inflammatory properties.
  • Neuroprotective agent.
  • Protects nerve cells from apoptosis and autophagy.
  • Inhibits expression of α-synuclein.
  • Provides myocardial protection.
  • Useful for research in neurological and cardiovascular diseases.