Learn More
Medchemexpress LLC Beta-asarone (standard) | 5273-86-9 | 208.25 | 5 MG
SDP
Catalog No. 5000415047
Encompass_Preferred
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent, which is the major ingredient of Acorus tatarinowii Schott. It can protect nerve cells from apoptosis and autophagy, inhibit expression of α-synuclein, and provides myocardial protection. Beta-asarone can be used in the study of neurological and cardiovascular diseases.
  • Orally active
  • BBB-penetrable anti-inflammatory and neuroprotective agent
  • Protects nerve cells from apoptosis and autophagy
  • Inhibits α-synuclein expression
  • Offers myocardial protection
  • Useful for studying neurological and cardiovascular diseases
Catalog No. 50-004-15047 Supplier Medchemexpress LLC Supplier No. HYN15015MG
Beta-asarone is an orally active and BBB-penetrable anti-inflammatory and neuroprotective agent, which is the major ingredient of Acorus tatarinowii Schott. It can protect nerve cells from apoptosis and autophagy, inhibit expression of α-synuclein, and provides myocardial protection. Beta-asarone can be used in the study of neurological and cardiovascular diseases.
  • Orally active
  • BBB-penetrable anti-inflammatory and neuroprotective agent
  • Protects nerve cells from apoptosis and autophagy
  • Inhibits α-synuclein expression
  • Offers myocardial protection
  • Useful for studying neurological and cardiovascular diseases