Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(beta,beta-Dimethylacryl)shikonin 90.0+%, TCI America™
SDP

Catalog No. D2118100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2118100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2118100MG Supplier TCI America Supplier No. D2118100MG

Chemical Identifiers

CAS 24502-79-2
Molecular Formula C21H22O6
Molecular Weight (g/mol) 370.401
MDL Number MFCD00143687
InChI Key BATBOVZTQBLKIL-QGZVFWFLSA-N
Synonym 5,8-Dihydroxy-2-[1-(beta,beta-dimethylacryloyloxy)-4-methyl-3-pentenyl]-1,4-naphthalenedione
PubChem CID 479499
ChEBI CHEBI:81071
IUPAC Name [(1R)-1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-enyl] 3-methylbut-2-enoate
SMILES CC(=CCC(C1=CC(=O)C2=C(C=CC(=C2C1=O)O)O)OC(=O)C=C(C)C)C

Specifications

Melting Point 103°C
Quantity 100 mg
Formula Weight 370.40
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (beta,beta-Dimethylacryl)shikonin
RTECSNumber EM9252400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.