Learn More
TARGETMOL CHEMICALS INC beta beta-Dimethylacrylalkanni
SDP
Catalog No. 502876995
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta,beta-Dimethylacrylalkannin is a natural product isolated from Arnebia nobilis Reichb.f, increases collagen and involucrin content in skin cells. Purity ≥95%
Catalog No. 50-287-6995 Supplier TARGETMOL CHEMICALS INC Supplier No. T570325MG
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta,beta-Dimethylacrylalkannin is a natural product isolated from Arnebia nobilis Reichb.f, increases collagen and involucrin content in skin cells. Purity ≥95%