Learn More
Research Products International Corp Beta Blocking Apron, Neoprene, 20 Inches Wide x 32 Inches Long, Black
SDP
Catalog No. 50213202
Encompass
This apron reduces beta emissions from 32P at normal working distances from the radiation source. Apron is fabricated from 1/8 inch thick black neoprene rubber and supplied with adjustable webbing straps and plastic buckles. Strap system places weight of apron on the shoulders instead of the neck.
This item is not returnable. View return policy
Catalog No. 50-213-202 Supplier Research Products International Corp Supplier No. BBA01
This apron reduces beta emissions from 32P at normal working distances from the radiation source. Apron is fabricated from 1/8 inch thick black neoprene rubber and supplied with adjustable webbing straps and plastic buckles. Strap system places weight of apron on the shoulders instead of the neck.