Learn More
Selleck Chemical LLC Beta Carotene S1767-50mg
SDP
Catalog No. 503110604
Encompass_Preferred
Beta Carotene( -Carotene) is an organic compound and classified as a terpenoid It is a precursor (inactive form) of vitamin A This product has poor solubility animal experiments are available cell experiments please choose carefully
Catalog No. 50-311-0604 Supplier Selleck Chemical LLC Supplier No. S176750MG
Beta Carotene( -Carotene) is an organic compound and classified as a terpenoid It is a precursor (inactive form) of vitamin A This product has poor solubility animal experiments are available cell experiments please choose carefully