Learn More
TARGETMOL CHEMICALS INC beta-Carotene 500MG
SDP
Catalog No. 502878749
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta-Carotene (Provitamin A) is a naturally-occurring retinol (vitamin A) precursor obtained from certain fruits and vegetables with potential antineoplastic and chemopreventive activities. As an anti-oxidant, beta carotene inhibits free-radical damage to DNA. Purity 98.99%
Catalog No. 50-287-8749 Supplier TARGETMOL CHEMICALS INC Supplier No. T1633500MG
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta-Carotene (Provitamin A) is a naturally-occurring retinol (vitamin A) precursor obtained from certain fruits and vegetables with potential antineoplastic and chemopreventive activities. As an anti-oxidant, beta carotene inhibits free-radical damage to DNA. Purity 98.99%